siRNA Details
| Gene Symbol: 4930509G22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015735.1 | - | 1 | 2082 | ||
| Gene Symbol: LOC105246071 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK134561.1 | - | 79 | |||

4930509G22Rik
Genomic Map