siRNA Details
| Gene Symbol: A230028O05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040374.1 | 3 | 1051 | |||
| GenBank mRNA | AK038535.1 | - | 3 | 1052 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A230028O05Rik| Gene Symbol: A230028O05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040374.1 | 3 | 1051 | |||
| GenBank mRNA | AK038535.1 | - | 3 | 1052 | ||