siRNA Details
| Gene Symbol: Gm26881 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043958.1 | - | 4 | 566 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm26881| Gene Symbol: Gm26881 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043958.1 | - | 4 | 566 | ||