siRNA Details
Gene Symbol: Gm16845 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040406.1 | 4 | 1124 | |||
NR_040407.1 | 3 | 1060 | ||||
GenBank mRNA | AK154427.1 | - | 3 | 1030 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm16845 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040406.1 | 4 | 1124 | |||
NR_040407.1 | 3 | 1060 | ||||
GenBank mRNA | AK154427.1 | - | 3 | 1030 |