siRNA Details
| Gene Symbol: Redrum | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040338.1 | 2 | 286 | |||
| GenBank mRNA | BY140310.1 | - | 2 | 266 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AI119445.1 | - | 2 | 254 | Chr. 18 - 54422327 - 54451636 [+] on Build GRCm38 | |

Redrum
Genomic Map