siRNA Details
| Gene Symbol: 2010015M23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008259.1 | - | 1 | 686 | ||
| AK041907.1 | - | 1 | 485 | |||
| AV065408.1 | - | 1 | 203 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CB192857.1 | - | 1 | 656 | Chr. 11 - 88046806 - 88047528 [-] on Build GRCm38 | |

2010015M23Rik
Genomic Map