siRNA Details
| Gene Symbol: 4930509E16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045735.1 | 2 | 118 | |||
| GenBank mRNA | AK015733.1 | - | 2 | 118 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715275.1 | - | 2 | 115 | Chr. 9 - 72518525 - 72531856 [-] on Build GRCm38 | |

4930509E16Rik
Genomic Map