siRNA Details
| Gene Symbol: Gm26710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133627.1 | - | 578 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ048224.1 | - | 2 | 578 | Chr. 10 - 80578647 - 80579496 [+] on Build GRCm38 | |

Gm26710
Genomic Map