siRNA Details
| Gene Symbol: 1110036E04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040713.1 | 3 | 514 | |||
| GenBank mRNA | AK004124.1 | - | 3 | 513 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY704366.1 | - | 3 | 509 | Chr. 9 - 64049833 - 64054100 [+] on Build GRCm38 | |

1110036E04Rik
Genomic Map