siRNA Details
| Gene Symbol: Pvt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003368.1 | 2 | 460 | |||
| GenBank mRNA | AK020628.1 | - | 2 | 437 | ||
| AK043790.1 | - | 2 | 462 | |||
| AK090048.1 | - | 397 | ||||
| AK144788.1 | - | 397 | ||||
| AK144854.1 | - | 420 | ||||
| AK169445.1 | - | 2 | 401 | |||

Pvt1
Genomic Map