siRNA Details
| Gene Symbol: 2610316D01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045172.1 | 5 | 449 | |||
| GenBank mRNA | AK012035.1 | - | 5 | 449 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610316D01Rik| Gene Symbol: 2610316D01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045172.1 | 5 | 449 | |||
| GenBank mRNA | AK012035.1 | - | 5 | 449 | ||