siRNA Details
| Gene Symbol: 1600020E01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC029726.1 | - | 700 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI415363.1 | - | 1 | 220 | Chr. 6 - 86526752 - 86564332 [+] on Build GRCm38 | |

1600020E01Rik
Genomic Map