siRNA Details
| Gene Symbol: C330046G13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040658.1 | 2, 3 | 139 | |||
| GenBank mRNA | AK082870.1 | - | 2, 3 | 140 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C330046G13Rik| Gene Symbol: C330046G13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040658.1 | 2, 3 | 139 | |||
| GenBank mRNA | AK082870.1 | - | 2, 3 | 140 | ||