siRNA Details
| Gene Symbol: 4930526L06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045317.1 | 3 | 442 | |||
| GenBank mRNA | AK015905.1 | - | 3 | 443 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930526L06Rik| Gene Symbol: 4930526L06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045317.1 | 3 | 442 | |||
| GenBank mRNA | AK015905.1 | - | 3 | 443 | ||