siRNA Details
| Gene Symbol: Gm19705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045324.1 | 3 | 194 | |||
| NR_045325.1 | 3 | 191 | ||||
| GenBank mRNA | AV450327.1 | - | 3 | 191 | ||
| BE447944.1 | - | 3 | 194 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm19705| Gene Symbol: Gm19705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045324.1 | 3 | 194 | |||
| NR_045325.1 | 3 | 191 | ||||
| GenBank mRNA | AV450327.1 | - | 3 | 191 | ||
| BE447944.1 | - | 3 | 194 | |||