siRNA Details
| Gene Symbol: Bcl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK145276.1 | - | 2 | 1502 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcl9| Gene Symbol: Bcl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK145276.1 | - | 2 | 1502 | ||