siRNA Details
| Gene Symbol: Gm16982 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040337.1 | 4 | 1007 | |||
| GenBank mRNA | AK016549.1 | - | 4 | 1008 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16982| Gene Symbol: Gm16982 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040337.1 | 4 | 1007 | |||
| GenBank mRNA | AK016549.1 | - | 4 | 1008 | ||