siRNA Details
| Gene Symbol: 9330159M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037982.1 | 5 | 526 | |||
| NR_037983.1 | 5 | 489 | ||||
| GenBank mRNA | AK034149.1 | - | 5 | 473 | ||
| AK040733.1 | - | 5 | 527 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9330159M07Rik| Gene Symbol: 9330159M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037982.1 | 5 | 526 | |||
| NR_037983.1 | 5 | 489 | ||||
| GenBank mRNA | AK034149.1 | - | 5 | 473 | ||
| AK040733.1 | - | 5 | 527 | |||