siRNA Details
| Gene Symbol: 9330159M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007093.1 | - | 716 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9330159M07Rik| Gene Symbol: 9330159M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007093.1 | - | 716 | |||