siRNA Details
| Gene Symbol: 2700038G22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045040.1 | 4 | 290 | |||
| NR_045042.1 | 4 | 271 | ||||
| GenBank mRNA | AK002390.1 | - | 4 | 290 | ||
| BG078826.2 | - | 3 | 263 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2700038G22Rik| Gene Symbol: 2700038G22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045040.1 | 4 | 290 | |||
| NR_045042.1 | 4 | 271 | ||||
| GenBank mRNA | AK002390.1 | - | 4 | 290 | ||
| BG078826.2 | - | 3 | 263 | |||