siRNA Details
| Gene Symbol: 4632428C04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033631.1 | 3 | 584 | |||
| GenBank mRNA | AK085759.1 | - | 3 | 586 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB661866.1 | - | 3 | 585 | Chr. 16 - 30008667 - 30019675 [+] on Build GRCm38 | |

4632428C04Rik
Genomic Map