siRNA Details
| Gene Symbol: Gm2449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132067.1 | - | 3 | 437 | ||
| AV160201.1 | - | 1 | 145 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712885.1 | - | 3 | 432 | Chr. 9 - 119892701 - 119894859 [-] on Build GRCm38 | |

Gm2449
Genomic Map