siRNA Details
| Gene Symbol: A330076H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015599.2 | 2 | 381 | |||
| GenBank mRNA | AK036297.1 | - | 2 | 382 | ||
| AK038438.1 | - | 435 | ||||
| AK039637.1 | - | 2 | 409 | |||
| AK080655.2 | - | 438 | ||||
| AK083195.1 | - | 2 | 434 | |||

A330076H08Rik
Genomic Map