siRNA Details
| Gene Symbol: 1500012K07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045814.1 | 3 | 1963 | |||
| GenBank mRNA | AK035406.1 | - | 3 | 1962 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1500012K07Rik| Gene Symbol: 1500012K07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045814.1 | 3 | 1963 | |||
| GenBank mRNA | AK035406.1 | - | 3 | 1962 | ||