siRNA Details
| Gene Symbol: A230107N01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039207.1 | - | 1 | 979 | ||
| AK053270.1 | - | 2 | 808 | |||
| BB183586.2 | - | 1 | 154 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A230107N01Rik| Gene Symbol: A230107N01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039207.1 | - | 1 | 979 | ||
| AK053270.1 | - | 2 | 808 | |||
| BB183586.2 | - | 1 | 154 | |||