siRNA Details
| Gene Symbol: Gm10516 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033536.1 | 3, 4 | 494 | |||
| GenBank mRNA | AK142593.1 | - | 3, 4 | 495 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10516| Gene Symbol: Gm10516 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033536.1 | 3, 4 | 494 | |||
| GenBank mRNA | AK142593.1 | - | 3, 4 | 495 | ||