siRNA Details
| Gene Symbol: 4931403G20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038172.1 | 4 | 1099 | |||
| NR_038173.1 | 3 | 951 | ||||
| GenBank mRNA | AK016427.1 | - | 4 | 1091 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4931403G20Rik| Gene Symbol: 4931403G20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038172.1 | 4 | 1099 | |||
| NR_038173.1 | 3 | 951 | ||||
| GenBank mRNA | AK016427.1 | - | 4 | 1091 | ||