siRNA Details
| Gene Symbol: Gm26671 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040865.1 | - | 4 | 775 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CB525728.1 | - | 4 | 661 | Chr. 3 - 57621259 - 57625788 [-] on Build GRCm38 | |

Gm26671
Genomic Map