siRNA Details
| Gene Symbol: C030034L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015499.2 | 1 | 24 | |||
| GenBank mRNA | AK030949.1 | - | 1 | 27 | ||
| AK037741.1 | - | 1 | 94 | |||
| AK039918.1 | - | 39 | ||||
| AK053975.1 | - | 1 | 39 | |||
| AK081263.1 | - | 35 | ||||

C030034L19Rik
Genomic Map