siRNA Details
| Gene Symbol: 1500026H17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK048993.1 | - | 3 | 1459 | ||
| Gene Symbol: AU040096 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139567.1 | - | 2 | 1278 | ||
| BG084238.1 | - | 1 | 87 | |||

1500026H17Rik
Genomic Map