siRNA Details
| Gene Symbol: 4930563E18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045379.1 | 1 | 337 | |||
| GenBank mRNA | AK016200.1 | - | 1 | 338 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930563E18Rik| Gene Symbol: 4930563E18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045379.1 | 1 | 337 | |||
| GenBank mRNA | AK016200.1 | - | 1 | 338 | ||