siRNA Details
| Gene Symbol: Mir155hg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY096003.1 | - | LIT1587 bic |
2 | 1103 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mir155hg| Gene Symbol: Mir155hg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY096003.1 | - | LIT1587 bic |
2 | 1103 | |