siRNA Details
| Gene Symbol: 1700030M09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045903.1 | 3 | 832 | |||
| GenBank mRNA | AK006561.1 | - | 3 | 833 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700030M09Rik| Gene Symbol: 1700030M09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045903.1 | 3 | 832 | |||
| GenBank mRNA | AK006561.1 | - | 3 | 833 | ||