siRNA Details
Gene Symbol: 1700030M09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045903.1 | 3 | 874 | |||
GenBank mRNA | AK006561.1 | - | 3 | 875 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700030M09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045903.1 | 3 | 874 | |||
GenBank mRNA | AK006561.1 | - | 3 | 875 |