siRNA Details
| Gene Symbol: AW047730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC072659.1 | - | 1343 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AW047730| Gene Symbol: AW047730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC072659.1 | - | 1343 | |||