siRNA Details
| Gene Symbol: Gm26782 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015096.1 | - | 4 | 908 | ||
| AK033525.1 | - | 772 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm26782| Gene Symbol: Gm26782 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015096.1 | - | 4 | 908 | ||
| AK033525.1 | - | 772 | ||||