siRNA Details
| Gene Symbol: LOC100862268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105029.1 | 4 | 1601 | |||
| NR_105030.1 | 3 | 1531 | ||||
| GenBank mRNA | AK138534.1 | - | 3 | 1532 | ||
| BB750343.1 | - | 1 | 212 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100862268| Gene Symbol: LOC100862268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105029.1 | 4 | 1601 | |||
| NR_105030.1 | 3 | 1531 | ||||
| GenBank mRNA | AK138534.1 | - | 3 | 1532 | ||
| BB750343.1 | - | 1 | 212 | |||