siRNA Details
| Gene Symbol: LOC100862268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105029.1 | 4 | 834 | |||
| NR_105030.1 | 3 | 764 | ||||
| GenBank mRNA | AK029402.1 | - | 4 | 801 | ||
| AK138534.1 | - | 3 | 765 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV251342.2 | - | 2 | 638 | Chr. 3 - 127166275 - 127167317 [+] on Build GRCm38 | |

LOC100862268
Genomic Map