siRNA Details
| Gene Symbol: 5830432E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015548.1 | 1 | 219 | |||
| GenBank mRNA | AK017962.1 | - | 1 | 220 | ||
| AK020025.1 | - | 1 | 217 | |||
| AK088640.1 | - | 1 | 190 | |||
| AK148045.1 | - | 188 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5830432E09Rik| Gene Symbol: 5830432E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015548.1 | 1 | 219 | |||
| GenBank mRNA | AK017962.1 | - | 1 | 220 | ||
| AK020025.1 | - | 1 | 217 | |||
| AK088640.1 | - | 1 | 190 | |||
| AK148045.1 | - | 188 | ||||