siRNA Details
| Gene Symbol: Gm2366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077006.1 | - | 2 | 3011 | ||
| AK133166.1 | - | 2 | 3011 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm2366| Gene Symbol: Gm2366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077006.1 | - | 2 | 3011 | ||
| AK133166.1 | - | 2 | 3011 | |||