siRNA Details
| Gene Symbol: 4933406C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_044986.1 | 2 | 397 | |||
| GenBank mRNA | AK016682.1 | - | 2 | 393 | ||
| AK041289.1 | - | 2 | 398 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933406C10Rik| Gene Symbol: 4933406C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_044986.1 | 2 | 397 | |||
| GenBank mRNA | AK016682.1 | - | 2 | 393 | ||
| AK041289.1 | - | 2 | 398 | |||