siRNA Details
| Gene Symbol: 3110045C21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040438.1 | 1 | 151 | |||
| GenBank mRNA | AK014032.1 | - | 1 | 109 | ||
| AK014177.1 | - | 1 | 152 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

3110045C21Rik| Gene Symbol: 3110045C21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040438.1 | 1 | 151 | |||
| GenBank mRNA | AK014032.1 | - | 1 | 109 | ||
| AK014177.1 | - | 1 | 152 | |||