siRNA Details
Gene Symbol: 3110045C21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040438.1 | 3 | 1054 | |||
GenBank mRNA | AK014032.1 | - | 3 | 992 | ||
AK014177.1 | - | 3 | 1055 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 3110045C21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040438.1 | 3 | 1054 | |||
GenBank mRNA | AK014032.1 | - | 3 | 992 | ||
AK014177.1 | - | 3 | 1055 |