siRNA Details
| Gene Symbol: B230110C06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020961.1 | - | 630 | |||
| AK080648.1 | - | 4 | 856 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY727002.1 | - | 3 | 626 | Chr. 14 - 15437626 - 15451834 [+] on Build GRCm38 | |

B230110C06Rik
Genomic Map