siRNA Details
| Gene Symbol: Jpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050201.1 | - | 1, 2 | 153 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV107747.2 | - | 1, 2 | 177 | Chr. X - 103493770 - 103503237 [+] on Build GRCm38 | |

Jpx
Genomic Map