siRNA Details
| Gene Symbol: C920006O11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040401.1 | 4 | 682 | |||
| GenBank mRNA | AK050586.1 | - | 4 | 683 | ||
| AK155758.1 | - | 2207 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C920006O11Rik| Gene Symbol: C920006O11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040401.1 | 4 | 682 | |||
| GenBank mRNA | AK050586.1 | - | 4 | 683 | ||
| AK155758.1 | - | 2207 | ||||