siRNA Details
| Gene Symbol: C920006O11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040401.1 | 3 | 320 | |||
| GenBank mRNA | AK036298.1 | - | 1 | 1112 | ||
| AK050586.1 | - | 3 | 321 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C920006O11Rik| Gene Symbol: C920006O11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040401.1 | 3 | 320 | |||
| GenBank mRNA | AK036298.1 | - | 1 | 1112 | ||
| AK050586.1 | - | 3 | 321 | |||