siRNA Details
| Gene Symbol: Cyp11a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042878.1 | - | 300 | |||
| AK137397.1 | - | 273 | ||||
| AK163559.1 | - | 312 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp11a1| Gene Symbol: Cyp11a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042878.1 | - | 300 | |||
| AK137397.1 | - | 273 | ||||
| AK163559.1 | - | 312 | ||||