siRNA Details
| Gene Symbol: Gm2716 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077166.1 | - | 8 | 1520 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV280686.2 | - | 4 | 608 | Chr. 8 - 88044898 - 88065341 [-] on Build GRCm38 | |

Gm2716
Genomic Map