siRNA Details
| Gene Symbol: Gm10532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045879.1 | 4 | 1811 | |||
| GenBank mRNA | AK133424.1 | - | 4 | 1812 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10532| Gene Symbol: Gm10532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045879.1 | 4 | 1811 | |||
| GenBank mRNA | AK133424.1 | - | 4 | 1812 | ||